Anti-TRIM13 Mouse Monoclonal Antibody [clone: 2A11]

Supplier: Abnova
H00010206-M01
10599-614EA 335 USD
10599-614
Anti-TRIM13 Mouse Monoclonal Antibody [clone: 2A11]
Antibodies
Type: Primary
Antigen: TRIM13
Clonality: Monoclonal
Clone: 2A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR