Anti-MPZL2 Mouse Monoclonal Antibody [clone: 2E10]

Supplier: Abnova
H00010205-M06
10593-680EA 335 USD
10593-680
Anti-MPZL2 Mouse Monoclonal Antibody [clone: 2E10]
Antibodies
Type: Primary
Antigen: MPZL2
Clonality: Monoclonal
Clone: 2E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR