Anti-NME6 Mouse Monoclonal Antibody [clone: 2A10]

Supplier: Abnova
H00010201-M07
10595-036EA 335 USD
10595-036
Anti-NME6 Mouse Monoclonal Antibody [clone: 2A10]
Antibodies
Type: Primary
Antigen: NME6
Clonality: Monoclonal
Clone: 2A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR