Anti-MPHOSPH6 Mouse Monoclonal Antibody [clone: 1D8-2A8]

Supplier: Abnova
H00010200-M01
10593-742EA 335 USD
10593-742
Anti-MPHOSPH6 Mouse Monoclonal Antibody [clone: 1D8-2A8]
Antibodies
Type: Primary
Antigen: MPHOSPH6
Clonality: Monoclonal
Clone: 1D8-2A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR