Anti-DHRS9 Mouse Monoclonal Antibody [clone: 3C6]

Supplier: Abnova
H00010170-M05
10583-906EA 335 USD
10583-906
Anti-DHRS9 Mouse Monoclonal Antibody [clone: 3C6]
Antibodies
Type: Primary
Antigen: DHRS9
Clonality: Monoclonal
Clone: 3C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR