Anti-CHST4 Mouse Monoclonal Antibody [clone: 4D7]

Supplier: Abnova
H00010164-M10
10582-226EA 335 USD
10582-226
Anti-CHST4 Mouse Monoclonal Antibody [clone: 4D7]
Antibodies
Type: Primary
Antigen: CHST4
Clonality: Monoclonal
Clone: 4D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR