Anti-WASF2 Mouse Monoclonal Antibody [clone: 1F7]

Supplier: Abnova
H00010163-M01
10605-726EA 335 USD
10605-726
Anti-WASF2 Mouse Monoclonal Antibody [clone: 1F7]
Antibodies
Type: Primary
Antigen: WASF2
Clonality: Monoclonal
Clone: 1F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR