Anti-G3BP1 Mouse Monoclonal Antibody [clone: 2F3]

Supplier: Abnova
H00010146-M01
10586-924EA 335 USD
10586-924
Anti-G3BP1 Mouse Monoclonal Antibody [clone: 2F3]
Antibodies
Type: Primary
Antigen: G3BP1 (GTPase activating protein (SH3 domain) binding protein 1)
Clonality: Monoclonal
Clone: 2F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR