Anti-CELA3A Rabbit Polyclonal Antibody

Supplier: Abnova
H00010136-D01
10584-876EA 376 USD
10584-876
Anti-CELA3A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CELA3A
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR