Anti-ZNF263 Mouse Monoclonal Antibody [clone: 2G6]

Supplier: Abnova
H00010127-M05
10606-404EA 335 USD
10606-404
Anti-ZNF263 Mouse Monoclonal Antibody [clone: 2G6]
Antibodies
Type: Primary
Antigen: ZNF263
Clonality: Monoclonal
Clone: 2G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR