Anti-ACTR1B Mouse Monoclonal Antibody [clone: 40]

Supplier: Abnova
H00010120-M05
10575-858EA 335 USD
10575-858
Anti-ACTR1B Mouse Monoclonal Antibody [clone: 40]
Antibodies
Type: Primary
Antigen: ACTR1B
Clonality: Monoclonal
Clone: 40
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR