Anti-SGK2 Mouse Monoclonal Antibody [clone: 1G11]

Supplier: Abnova
H00010110-M13
10600-820EA 335 USD
10600-820
Anti-SGK2 Mouse Monoclonal Antibody [clone: 1G11]
Antibodies
Type: Primary
Antigen: SGK2
Clonality: Monoclonal
Clone: 1G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR