Anti-SGK2 Mouse Monoclonal Antibody [clone: 3D2]

Supplier: Abnova
H00010110-M09
10601-244EA 335 USD
10601-244
Anti-SGK2 Mouse Monoclonal Antibody [clone: 3D2]
Antibodies
Type: Primary
Antigen: SGK2
Clonality: Monoclonal
Clone: 3D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR