Anti-SGK2 Mouse Monoclonal Antibody [clone: 4B12]

Supplier: Abnova
H00010110-M04
10571-094EA 335 USD
10571-094
Anti-SGK2 Mouse Monoclonal Antibody [clone: 4B12]
Antibodies
Type: Primary
Antigen: SGK2
Clonality: Monoclonal
Clone: 4B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR