Anti-SGK2 Mouse Monoclonal Antibody [clone: 1A7]

Supplier: Abnova
H00010110-M02
10600-824EA 335 USD
10600-824
Anti-SGK2 Mouse Monoclonal Antibody [clone: 1A7]
Antibodies
Type: Primary
Antigen: SGK2
Clonality: Monoclonal
Clone: 1A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR