Anti-CTDSP2 Mouse Monoclonal Antibody [clone: 3F11]

Supplier: Abnova
H00010106-M02A
10624-142EA 335 USD
10624-142
Anti-CTDSP2 Mouse Monoclonal Antibody [clone: 3F11]
Antibodies
Type: Primary
Antigen: CTDSP2
Clonality: Monoclonal
Clone: 3F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR