Anti-TSPAN1 Mouse Polyclonal Antibody

Supplier: Abnova
H00010103-A01
10573-114EA 243 USD
10573-114
Anti-TSPAN1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TSPAN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR