Anti-ACTR3 Mouse Monoclonal Antibody [clone: 2B6]

Supplier: Abnova
H00010096-M02
10576-176EA 335 USD
10576-176
Anti-ACTR3 Mouse Monoclonal Antibody [clone: 2B6]
Antibodies
Type: Primary
Antigen: ACTR3
Clonality: Monoclonal
Clone: 2B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR