Anti-COL4A3BP Mouse Polyclonal Antibody

Supplier: Abnova
H00010087-A01
10553-894EA 243 USD
10553-894
Anti-COL4A3BP Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: COL4A3BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR