Anti-PQBP1 Mouse Monoclonal Antibody [clone: 3H7]

Supplier: Abnova
H00010084-M15
10597-946EA 335 USD
10597-946
Anti-PQBP1 Mouse Monoclonal Antibody [clone: 3H7]
Antibodies
Type: Primary
Antigen: PQBP1
Clonality: Monoclonal
Clone: 3H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR