Anti-DPP3 Mouse Monoclonal Antibody [clone: 4D3]

Supplier: Abnova
H00010072-M01A
10624-190EA 335 USD
10624-190
Anti-DPP3 Mouse Monoclonal Antibody [clone: 4D3]
Antibodies
Type: Primary
Antigen: DPP3
Clonality: Monoclonal
Clone: 4D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR