Anti-ABCB6 Mouse Monoclonal Antibody [clone: 3E1]

Supplier: Abnova
H00010058-M01
10575-956EA 335 USD
10575-956
Anti-ABCB6 Mouse Monoclonal Antibody [clone: 3E1]
Antibodies
Type: Primary
Antigen: ABCB6
Clonality: Monoclonal
Clone: 3E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR