Anti-SAE1 Mouse Monoclonal Antibody [clone: 1G4-1G5]

Supplier: Abnova
H00010055-M01
10599-278EA 335 USD
10599-278
Anti-SAE1 Mouse Monoclonal Antibody [clone: 1G4-1G5]
Antibodies
Type: Primary
Antigen: SAE1
Clonality: Monoclonal
Clone: 1G4-1G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR