Anti-GJC1 Mouse Polyclonal Antibody

Supplier: Abnova
H00010052-A01
10557-500EA 243 USD
10557-500
Anti-GJC1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GJC1 (gap junction protein, gamma 1, 45kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR