Anti-SMC4 Mouse Polyclonal Antibody

Supplier: Abnova
H00010051-A01
10571-648EA 243 USD
10571-648
Anti-SMC4 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SMC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR