Anti-DNAJB6 Mouse Polyclonal Antibody

Supplier: Abnova
H00010049-A01
10555-234EA 243 USD
10555-234
Anti-DNAJB6 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DNAJB6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR