Anti-SH2D3C Mouse Monoclonal Antibody [clone: 2000]

Supplier: Abnova
H00010044-M02
10601-298EA 335 USD
10601-298
Anti-SH2D3C Mouse Monoclonal Antibody [clone: 2000]
Antibodies
Type: Primary
Antigen: SH2D3C
Clonality: Monoclonal
Clone: 2000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR