Anti-SH2D3C Mouse Monoclonal Antibody [clone: 3B2]

Supplier: Abnova
H00010044-M01
10601-296EA 335 USD
10601-296
Anti-SH2D3C Mouse Monoclonal Antibody [clone: 3B2]
Antibodies
Type: Primary
Antigen: SH2D3C
Clonality: Monoclonal
Clone: 3B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR