Anti-TOM1 Mouse Monoclonal Antibody [clone: 5A3]

Supplier: Abnova
H00010043-M01
10604-384EA 335 USD
10604-384
Anti-TOM1 Mouse Monoclonal Antibody [clone: 5A3]
Antibodies
Type: Primary
Antigen: TOM1
Clonality: Monoclonal
Clone: 5A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR