Anti-CHAF1A Mouse Monoclonal Antibody [clone: 1C2]

Supplier: Abnova
H00010036-M02
10582-096EA 335 USD
10582-096
Anti-CHAF1A Mouse Monoclonal Antibody [clone: 1C2]
Antibodies
Type: Primary
Antigen: CHAF1A
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR