Anti-MED16 Mouse Monoclonal Antibody [clone: 2B7]

Supplier: Abnova
H00010025-M02
10603-766EA 335 USD
10603-766
Anti-MED16 Mouse Monoclonal Antibody [clone: 2B7]
Antibodies
Type: Primary
Antigen: MED16
Clonality: Monoclonal
Clone: 2B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR