Anti-GNE Rabbit Polyclonal Antibody

Supplier: Abnova
H00010020-D01P
10587-602EA 376 USD
10587-602
Anti-GNE Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GNE (glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR