Anti-PDCD6IP Mouse Monoclonal Antibody [clone: 3C4]

Supplier: Abnova
H00010015-M01
10596-394EA 335 USD
10596-394
Anti-PDCD6IP Mouse Monoclonal Antibody [clone: 3C4]
Antibodies
Type: Primary
Antigen: PDCD6IP
Clonality: Monoclonal
Clone: 3C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR