Anti-GNPDA1 Mouse Polyclonal Antibody

Supplier: Abnova
H00010007-A01
10560-934EA 243 USD
10560-934
Anti-GNPDA1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GNPDA1 (glucosamine-6-phosphate deaminase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR