Anti-AKT3 Mouse Monoclonal Antibody [clone: 60]

Supplier: Abnova
H00010000-M02
10576-612EA 335 USD
10576-612
Anti-AKT3 Mouse Monoclonal Antibody [clone: 60]
Antibodies
Type: Primary
Antigen: AKT3
Clonality: Monoclonal
Clone: 60
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR