Anti-PTBP3 Mouse Monoclonal Antibody [clone: 4C9]

Supplier: Abnova
H00009991-M01
10599-606EA 335 USD
10599-606
Anti-PTBP3 Mouse Monoclonal Antibody [clone: 4C9]
Antibodies
Type: Primary
Antigen: PTBP3
Clonality: Monoclonal
Clone: 4C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR