Anti-DMTF1 Mouse Monoclonal Antibody [clone: 5C6]

Supplier: Abnova
H00009988-M03
10584-156EA 335 USD
10584-156
Anti-DMTF1 Mouse Monoclonal Antibody [clone: 5C6]
Antibodies
Type: Primary
Antigen: DMTF1
Clonality: Monoclonal
Clone: 5C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR