Anti-CCS Mouse Monoclonal Antibody [clone: 100]

Supplier: Abnova
H00009973-M03
10578-986EA 335 USD
10578-986
Anti-CCS Mouse Monoclonal Antibody [clone: 100]
Antibodies
Type: Primary
Antigen: CCS
Clonality: Monoclonal
Clone: 100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR