Anti-CCS Mouse Polyclonal Antibody

Supplier: Abnova
H00009973-B01
10552-778EA 335 USD
10552-778
Anti-CCS Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CCS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR