Anti-HS3ST3A1 Rabbit Polyclonal Antibody

Supplier: Abnova
H00009955-D01P
10589-320EA 376 USD
10589-320
Anti-HS3ST3A1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HS3ST3A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR