Anti-OXSR1 Mouse Monoclonal Antibody [clone: 2A5]

Supplier: Abnova
H00009943-M20
10595-822EA 335 USD
10595-822
Anti-OXSR1 Mouse Monoclonal Antibody [clone: 2A5]
Antibodies
Type: Primary
Antigen: OXSR1
Clonality: Monoclonal
Clone: 2A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR