Anti-EXOG Rabbit Polyclonal Antibody

Supplier: Abnova
H00009941-D01P
10585-002EA 376 USD
10585-002
Anti-EXOG Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EXOG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR