Anti-PUM3 Rabbit Polyclonal Antibody

Supplier: Abnova
H00009933-D01
10591-016EA 376 USD
10591-016
Anti-PUM3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PUM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR