Anti-HELZ Mouse Monoclonal Antibody [clone: 1C11]

Supplier: Abnova
H00009931-M04
10588-576EA 335 USD
10588-576
Anti-HELZ Mouse Monoclonal Antibody [clone: 1C11]
Antibodies
Type: Primary
Antigen: HELZ
Clonality: Monoclonal
Clone: 1C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR