Anti-HELZ Mouse Monoclonal Antibody [clone: 5B2]

Supplier: Abnova
H00009931-M02
10588-578EA 335 USD
10588-578
Anti-HELZ Mouse Monoclonal Antibody [clone: 5B2]
Antibodies
Type: Primary
Antigen: HELZ
Clonality: Monoclonal
Clone: 5B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR