Anti-MFN2 Mouse Monoclonal Antibody [clone: 6A8]

Supplier: Abnova
H00009927-M01
10593-276EA 335 USD
10593-276
Anti-MFN2 Mouse Monoclonal Antibody [clone: 6A8]
Antibodies
Type: Primary
Antigen: MFN2
Clonality: Monoclonal
Clone: 6A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR