Anti-PAN2 Mouse Monoclonal Antibody [clone: 6A7]

Supplier: Abnova
H00009924-M01
10605-512EA 335 USD
10605-512
Anti-PAN2 Mouse Monoclonal Antibody [clone: 6A7]
Antibodies
Type: Primary
Antigen: PAN2
Clonality: Monoclonal
Clone: 6A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR