Anti-NCAPD2 Mouse Monoclonal Antibody [clone: 4C9]

Supplier: Abnova
H00009918-M02
10582-462EA 335 USD
10582-462
Anti-NCAPD2 Mouse Monoclonal Antibody [clone: 4C9]
Antibodies
Type: Primary
Antigen: NCAPD2
Clonality: Monoclonal
Clone: 4C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR