Anti-MAGI2 Mouse Monoclonal Antibody [clone: 6F5]

Supplier: Abnova
H00009863-M02
10580-870EA 335 USD
10580-870
Anti-MAGI2 Mouse Monoclonal Antibody [clone: 6F5]
Antibodies
Type: Primary
Antigen: MAGI2
Clonality: Monoclonal
Clone: 6F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR