Anti-FARP2 Mouse Polyclonal Antibody

Supplier: Abnova
H00009855-A01
10556-362EA 243 USD
10556-362
Anti-FARP2 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FARP2 (FERM, RhoGEF and pleckstrin domain protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR